Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Phenoxyacetonitrile

Phenoxyacetonitrile

Glass bottle for solids, 25 g
Codice catalogo ACTIAS21875G25
Marketsource
Nome del fornitore:   ACTIVATE SCIENTIFIC
Phenoxyacetonitrile
Codice catalogo ACTIAS21875G25
Nome del fornitore:  ACTIVATE SCIENTIFIC
Marketsource
Numero CAS:  3598-14-9
Phenoxyacetonitrile

Glass bottle for solids, 25 g

Numero CAS:  3598-14-9
Price
Richiedi prezzo
Log in per disponibilità
1 * 25 g
Product Details & Documents
  • Numero CAS:3598-14-9
  • Formula:C₈H₇NO
  • Numero MDL:MFCD00052017
  • Molecular Weight:133.15 g/mol
  • UN:3276
  • ADR:6.1,III
Specifiche

Specifiche

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.