Combat shorts, S790
Nome del fornitore: Portwest
Informazioni su questo articolo
Made of Kingsmill fabric (65% polyester / 35% cotton), these work shorts are very tough and durable.
- Two side pockets, two back patch pockets, rule pocket
- Thigh pocket, Velcro® fastened, with mobile phone pocket
- Half elasticated waistband
Weight: 210 g/m²
3 varianti
Product Details & Documents
Made of Kingsmill fabric (65% polyester / 35% cotton), these work shorts are very tough and durable.
- Two side pockets, two back patch pockets, rule pocket
- Thigh pocket, Velcro® fastened, with mobile phone pocket
- Half elasticated waistband
Weight: 210 g/m²
Specifications for Product Family
Specifiche
- Catalog No:
- 113-7564
- 113-7563
- 113-7562
- Modello:
- S790
- S790
- S790
- Applicazioni per abiti:
- General Purpose|Industrial
- General Purpose|Industrial
- General Purpose|Industrial
- Genere:
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Sleeve or leg length:
- Short
- Short
- Short
- Tessuto:
- Kingsmill fabric (65% polyester, 35% cotton)
- Kingsmill fabric (65% polyester, 35% cotton)
- Kingsmill fabric (65% polyester, 35% cotton)
- Peso:
- 210 g/m²
- 210 g/m²
- 210 g/m²
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.113-7564ModelloS790Applicazioni per abitiGeneral Purpose|IndustrialGenerePer l'uomoSleeve or leg lengthShortTessutoKingsmill fabric (65% polyester, 35% cotton)Peso210 g/m² - Specifiche:
More
Specifications
Cat. No.113-7563ModelloS790Applicazioni per abitiGeneral Purpose|IndustrialGenerePer l'uomoSleeve or leg lengthShortTessutoKingsmill fabric (65% polyester, 35% cotton)Peso210 g/m² - Specifiche:
More
Specifications
Cat. No.113-7562ModelloS790Applicazioni per abitiGeneral Purpose|IndustrialGenerePer l'uomoSleeve or leg lengthShortTessutoKingsmill fabric (65% polyester, 35% cotton)Peso210 g/m²