Overalls with bib and braces, for food industries, Killybeg 6639
Supplier: SIOEN
Certificates
About this item
These reversible overalls are made of Flexothane® Kleen fabric consisting of 100% polyamide knitting 6.6 with PU coating. This material is durable, lightweight and most suitable for food contact.
- Waterproof and windproof
- Elastic braces with buckles on front and back
- Legs adjustable by press studs
- Washable at 95 °C
Weight: 170 g/m²
Certifications: Tested according to CE Directive 1935/2004 and ISO 10993-5 (1999).
EN 14605 (type PB/4), EN 343 (3-1)
EN 14605 (type PB/4), EN 343 (3-1)
Specifications
- Catalog No:
- 113-2224
- 113-2225
- 113-7163
- 113-2223
- 113-7165
- 113-7164
- 113-7167
- 113-7166
- 113-2227
- 113-2226
- 113-7168
- 113-2228
- Modello:
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- 6639 Killybeg
- Applicazioni per abiti:
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Tessuto:
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- 100% polyamide knitting 6.6 with PU coating
- Peso:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²