Capillary columns, VF-DA
Temp. limit -60 to 325/350 °C, Capillare, VF-DA, 0,20 mm, 12 m
Catalog # VARICP8964
Supplier: VARIAN
Jump to:
Product Details & Documents
VF-DA is a unique, guaranteed low bleed FactorFour GC column for drugs of abuse confirmation testing. The VF-DA column has high recovery for trace components and excellent resistance to direct methanol injections.
- Cross-linked and bonded to extend column lifetime
- Minimal bleed to improve detection limits and productivity
- High recovery of trace components to deliver accurate results
Specifications
- Formato della colonna:Capillare
- Ø int.:0,20 mm
- Descrizione:Temp. limit -60 to 325/350 °C
- Lunghezza:12 m
- Fase:VF-DA