Desiccators, MOBILEX type with procelain plate, DURAN®
Nome del fornitore: DWK Life Sciences
Certificati
Informazioni su questo articolo
DURAN® clear borosilicate glass 3.3, vacuum desiccator with Mobilex screw thread in the lid with stopcock. With desiccator base with flat flange, no outlet, PBT screw cap with pierced aperture GL 32.
Delivery: Supplied with porclain plate and silicone sealing ring with bonded PTFE face
Frequently Bought Together