Measuring cylinders, conical
Nome del fornitore: VITLAB
Certificati
Informazioni su questo articolo
PP, translucent.
- Good chemical resistance
- Autoclavable to +121 °C
- Liquid volume is easy to read
Ideal for a wide range of applications. With moulded graduations. Broad base for stability.
Frequently Bought Together