Emergency blanket
Emergency blanket, disposable, 1600×2100 mm
Codice catalogo 118-0252
Nome del fornitore: ORKLA CARE
Vai a
Product Details & Documents
This blanket, made of polyester film coated with aluminium on one side, is intended for use in a first aid situation. It helps prevent hypothermia, assists in prevention of post-accident shocks, and protects the injured person from wind and moisture.
Specifiche
- Descrizione:Emergency blanket, disposable
- L×L:1600×2100 mm