Orders LinkContactUsLinkComponent
Eyewash solutions in portable case
Eyewash solutions in portable case
Wall bracket for portable case, green metallic, 215×90×125 mm
Catalog # 118-0253
Supplier:   ORKLA CARE
Eyewash solutions in portable case
Catalog # 118-0253
Supplier:  ORKLA CARE
Eyewash solutions in portable case
Eyewash solutions in portable case
Wall bracket for portable case, green metallic, 215×90×125 mm
Price
SFr. 45.00
Log in for availability
1 * 1 ST
Product Details & Documents
This plastic, case with transparent lid and handle, contains five bottles of Cederroth eyewash solution. The solution is intended for washing away particles and dangerous liquids, like alkali or acids, from an eye in an emergency situation in the workplace.
  • Portable case is ideal for mobile workplaces and for continuous irrigation during transport to the hospital
  • Buffered eyewash has a neutralising effect on splashes from alkalis and acids
  • Box made of green polystyrene, lid made of transparent ABS
  • Case can be wall mounted by using the wall bracket
Specifications
  • Descrizione:
    Wall bracket for portable case, green metallic, 215×90×125 mm
  • Seq:
    02