Polymethylmethacrylate standards, GPC/SEC columns calibration
Nome del fornitore: VARIAN
Polymethylmethacrylate standards, GPC/SEC columns calibration
Nome del fornitore: VARIAN
Certificates
Informazioni su questo articolo
Polymethylmethacrylate (PMMA) standards are extremely versatile as they can be used for organic GPC with a wide range of medium polarity eluents, such as tetrahydrofuran, toluene, methyl ethyl ketone and ethyl acetate.
- Many solvent options increase applicability
- Stringent quality control improves performance
- Proprietary manufacturing methods ensure consistent supply
43 varianti
Product Details & Documents
Polymethylmethacrylate (PMMA) standards are extremely versatile as they can be used for organic GPC with a wide range of medium polarity eluents, such as tetrahydrofuran, toluene, methyl ethyl ketone and ethyl acetate.
- Many solvent options increase applicability
- Stringent quality control improves performance
- Proprietary manufacturing methods ensure consistent supply
Specifications for Product Family
Specifiche
Product Family Options
Calibration kit
Informazioni sul prodotto
- Pk InfoConfDisponibilitàPrice
- VARIPL2020-0100Calibration kit, M-L-10, 10×0,5 g, nominal Mp 600-50000 g/molSpecifiche:
More
Specifications
Cat. No.VARIPL2020-0100
Individual polymer molecular weight
Informazioni sul prodotto
- Pk InfoConfDisponibilitàPrice
- VARIPL2022-2001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,19, nominal Mp 500 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,19, nominal Mp 500 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-2001 - VARIPL2022-2005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,19, nominal Mp 500 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,19, nominal Mp 500 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-2005 - VARIPL2022-3001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,26, nominal Mp 1,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,26, nominal Mp 1,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-3001 - VARIPL2022-3005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,26, nominal Mp 1,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,26, nominal Mp 1,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-3005 - VARIPL2022-5001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 2,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 2,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-5001 - VARIPL2022-5005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 2,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 2,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-5005 - VARIPL2022-6001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 3,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 3,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-6001 - VARIPL2022-6005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 3,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 3,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-6005 - VARIPL2022-7001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 5,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 5,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-7001 - VARIPL2022-7005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 5,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 5,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-7005 - VARIPL2022-8001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 7,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 7,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-8001 - VARIPL2022-9001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 10,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 10,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-9001 - VARIPL2022-9005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 10,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 10,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2022-9005 - VARIPL2023-0001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 13,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 13,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-0001 - VARIPL2023-1001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 20,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 20,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-1001 - VARIPL2023-1005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 20,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 20,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-1005 - VARIPL2023-2001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 30,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 30,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-2001 - VARIPL2023-3001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 50,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 50,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-3001 - VARIPL2023-3005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 50,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 50,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-3005 - VARIPL2023-4001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 70,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 70,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-4001 - VARIPL2023-4005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 70,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 70,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-4005 - VARIPL2023-5001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 100,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 100,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-5001 - VARIPL2023-5005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 100,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 100,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-5005 - VARIPL2023-5010Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 100,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 100,000 Da.10 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-5010 - VARIPL2023-6001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,05, nominal Mp 130,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,05, nominal Mp 130,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-6001 - VARIPL2023-7001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 200,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 200,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-7001 - VARIPL2023-7005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 200,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 200,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-7005 - VARIPL2023-7010Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 200,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 200,000 Da.10 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-7010 - VARIPL2023-8001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 300,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 300,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-8001 - VARIPL2023-8005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 300,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,02, nominal Mp 300,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-8005 - VARIPL2023-9001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,06, nominal Mp 500,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,06, nominal Mp 500,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-9001 - VARIPL2023-9005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,06, nominal Mp 500,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,06, nominal Mp 500,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-9005 - VARIPL2023-9010Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,06, nominal Mp 500,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,06, nominal Mp 500,000 Da.10 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2023-9010 - VARIPL2024-0001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 700,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 700,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-0001 - VARIPL2024-0005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 700,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 700,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-0005 - VARIPL2024-0010Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 700,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 700,000 Da.10 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-0010 - VARIPL2024-1001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,000,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,000,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-1001 - VARIPL2024-1005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,000,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,000,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-1005 - VARIPL2024-1010Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,000,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,000,000 Da.10 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-1010 - VARIPL2024-2001Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da.1 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-2001 - VARIPL2024-2005Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da.5 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-2005 - VARIPL2024-2010Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da.Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da.10 g1Log in per disponibilità1 * 1 STSpecifiche:
More
Specifications
Cat. No.VARIPL2024-2010