Strabismus scissors, curved
Strabismus, curved, blunt/blunt, 110 mm
Codice catalogo MURR1913/10
Nome del fornitore: MURRAY S
Vai a
Product Details & Documents
These scissors are an ophthalmic dissecting instrument and are primarily used during procedures for the treatment of Strabismus.
Specifiche
- Tipo:Strabismus, curved, blunt/blunt
- Lunghezza:110 mm