Accessories for 2D barcode reader, VisionMate® High Speed
1D reader adapter, for reading and integrating codes on the sides of racks into file names
Codice catalogo MATRAB-1860
Nome del fornitore: THERMO MATRIX TECHNOLOGIES
Accessories for 2D barcode reader, VisionMate® High Speed
Codice catalogo MATRAB-1860
Nome del fornitore: THERMO MATRIX TECHNOLOGIES
Vai a
Product Details & Documents
Specifiche
- Descrizione:1D reader adapter, for reading and integrating codes on the sides of racks into file names
- Cat. No.:MATRAB-1860
- Peso:500 g
- Interfaccia:USB
- L×P×A:57×101×133 mm