ContactUsLinkComponent
RACK 5X96 3.0MM SHC Bruker caps are crafted from high-quality Orgalloy® plastic, offering excellent resistance to a variety of chemicals and NMR solvents such as CDCl3, MeOD, and DMSO. However, it is advisable to prevent direct contact between the sa 1 * 1 ST
Référence BRUKZ168407
Fournisseur:  BRUKER DALTRONIK
New Product
RACK 5X96 3.0MM SHC Bruker caps are crafted from high-quality Orgalloy® plastic, offering excellent resistance to a variety of chemicals and NMR solvents such as CDCl3, MeOD, and DMSO. However, it is advisable to prevent direct contact between the sa 1 * 1 ST
Référence BRUKZ168407
Fournisseur:  BRUKER DALTRONIK