ContactUsLinkComponent
CUSTOM Box of 96 PhyTip® columns (200 uL) containing 5 uL CaptureSelect IgA XL affinity matrix #2943972005 (Thermo Fisher) resin per column 1 * 96 ST
Référence MSPPOFPTR-92-05-QV
CUSTOM Box of 96 PhyTip® columns (200 uL) containing 5 uL CaptureSelect IgA XL affinity matrix #2943972005 (Thermo Fisher) resin per column 1 * 96 ST
Référence MSPPOFPTR-92-05-QV
Frequently Bought Together