Contact Plates, Gamma-Irradiated, Individually Packed
N° de catalogue LIOF15330
Fournisseur: LIOFILCHEM
New Product
Numéro CAS:
Spécifications
- Désignation:Tryptic Soy Agar + 0,1% Penase + Neutralising, single wrapped
- Seq:11
Spécifications
À propos de cet article
Contact plates are used for monitoring surfaces, air, and personnel inside cleanrooms and sterile areas. The peelable blister pack ensures the highest quality and protection standards for the plates and reduces waste.
Individual plate packaging reduces waste by keeping unused plates valid until shelf life ends
- Transparent packaging allows easy plate examination before opening
- Drastically reduces the dehydration of culture media during the shelf life period
- Shelf life up to 9 months
- Storage at ambient temperature for most items in its range
- Lowers contamination risks in controlled environments
Available as gamma-irradiated, triple-enveloped plates, suitable for restricted areas such as isolators and cleanrooms.
Certifications:
In compliance with European Pharmacopoeia, United States Pharmacopeia, and Japanese Pharmacopoeia.
Frequently Bought Together