ContactUsLinkComponent
Micro to Nano|Micro-Tec TA5 angled ultra-fine tungsten needle probe, Ø0.52mm with 0.6um tip, 25mm long 1 * 1 ST
  MSPP52-001045
Micro to Nano|Micro-Tec TA5 angled ultra-fine tungsten needle probe, Ø0.52mm with 0.6um tip, 25mm long 1 * 1 ST
  MSPP52-001045
Frequently Bought Together