with controller Pilot ONE Heat Exchanger Unit Temperature range: (3) ... (95) C Cooling power at 20C: 5 kW max. delivery pressure 4 bar max. delivery 30 l/min Pump connection: G3/4 male Power supply: 230V 1 50/60Hz Thermal fluid- primary circuit 1 * 1 items
Référence HUBEEVA14029
Fournisseur: HUBER