,,5 components: Bromides (Br-) 125mg/l Fluorides (F-) 125mg/l Nitrates (NO3-) 125mg/l Sulphates (SO42-) 500mg/l Chlorides (Cl-) 1250mg/l in Water ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory 1 * 100 mL
N° de catalogue CMPLRM116542L1
Fournisseur: CUSTOM MADE CHEMICALS LAB