,,13 components: Ca 1000mg/l Al 1000mg/l Li 1000mg/l Ni 1000mg/l Mg 1000mg/l Fe 1000mg/l As 100mg/l Cu 100mg/l Sb 100mg/l V 100mg/l Pb 100mg/l Cd 100mg/l Co 100mg/l in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 1 * 500 mL
N° de catalogue CMPLRM115955L5
Fournisseur: CUSTOM MADE CHEMICALS LAB