ContactUsLinkComponent
Bluetooth 5.1 EDR/2.1 BLE/V4.2, NFC (Not Available for JP) , Standard Range 2D Imager (SE4750SR) , 4GB RAM and 64GB ROM (UFS) , 16M Pixels Autofocus Camera , 25 Key Numeric Keypad ,USB Snap-On Kit EU Adapt 1 * 1 ST
Référence MSPPCLRK26-11
Bluetooth 5.1 EDR/2.1 BLE/V4.2, NFC (Not Available for JP) , Standard Range 2D Imager (SE4750SR) , 4GB RAM and 64GB ROM (UFS) , 16M Pixels Autofocus Camera , 25 Key Numeric Keypad ,USB Snap-On Kit EU Adapt 1 * 1 ST
Référence MSPPCLRK26-11
Frequently Bought Together