30 kDa Regenerated Cellulose µPULSE TFF system consumable chip (can be used for ~300mL of concentration this can be across multip le uses if cleaned and stored properly). Pack of 5. 1 * 5 ST
Référence MSPP809390B
Fournisseur: MARKET SOURCE PART PROCESS
Frequently Bought Together