ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 10 mL
N° de catalogue AATB22870
Fournisseur: AAT Bioquest
ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
N° de catalogue AATB22870
Fournisseur: AAT Bioquest
Numéro CAS:
Aller à
Product Details & Documents
The ReadiUse™ Annexin Binding Buffer *Optimised for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.
- Provided as a 10X concentrate
- Saves time
- pH of 7.4
Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.
Caution: For research use only.
Spécifications
Spécifications
- Cdt:10 mL
- Désignation:ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry



