ContactUsLinkComponent
Anti-His Antibody Terbium Cryptate Conjugation Service-1 mg volume with 1 week turnaround priority. The customer provides BSA and other amino acids free anti-His antibody in PBS, with a required concentration >/= 5 mg/mL. Expected yield of this servi 1 * 1 mg
Référence AATBQ240624B01-1MG
Fournisseur:  AAT Bioquest
New Product
Anti-His Antibody Terbium Cryptate Conjugation Service-1 mg volume with 1 week turnaround priority. The customer provides BSA and other amino acids free anti-His antibody in PBS, with a required concentration >/= 5 mg/mL. Expected yield of this servi 1 * 1 mg
Référence AATBQ240624B01-1MG
Fournisseur:  AAT Bioquest