,,12 components: Zn 9850mg/l Pb 200mg/l Fe 20mg/l Al 10mg/l Cu 10mg/l Sn 60mg/l Cd 10mg/l Ni 10mg/l Mn 10mg/l Mg 10mg/l Ti 10mg/l Bi 20mg/l in HCl 10% HNO3 5% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months.,,,, ,, 1 * 100 mL
N° de catalogue CMPLRM108737L1.10
Fournisseur: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together