Membrane potential detection kit, MITO-ID®
MITO-ID® membrane potential detection kit, 100 Tests
N° de catalogue ENZOENZ51018K100
Fournisseur: Enzo Life Sciences
Membrane potential detection kit, MITO-ID®
N° de catalogue ENZOENZ51018K100
Fournisseur: Enzo Life Sciences
Numéro CAS:
Aller à
Product Details & Documents
MITO-ID® membrane potential detection kit monitors energetic status using a simple mix-and-read, no-wash protocol. It measures mitochondrial membrane potential with a cationic dye that fluoresces either green or orange depending upon membrane potential status.
- Highly sensitive detection of mitochondrial membrane potential decline
- Aids in the drug research process in preclinical drug safety assessment (ADME-Tox)
- Suitable for chemical/environmental toxicity screening and characterisation
- True mix-and-read homogeneous assay for live cells
- Optimized for the measurement of mitochondria membrane potential (MMP) and cell viability by fluorescence microscopy and flow cytometry
Cell-based assays for evaluating the functional status of mitochondria are useful tools for elucidating the role of mitochondrial activity in drug-induced toxicity, the apoptosis cascade and other cellular and biochemical processes.
: Kit contains MITO-ID® MP detection reagent, necrosis detection reagent, CCCP control, 10X assay buffer 1 and 50X assay buffer 2.
Spécifications
Spécifications
- Cdt:100 Tests
- Désignation:MITO-ID® membrane potential detection kit