DNA isolation kit, EPIXTRACT®
EPIXTRACT® DNA isolation kit, 1 KIT
N° de catalogue ENZOENZ450180100
Fournisseur: Enzo Life Sciences
DNA isolation kit, EPIXTRACT®
N° de catalogue ENZOENZ450180100
Fournisseur: Enzo Life Sciences
Numéro CAS:
Aller à
Product Details & Documents
EPIXTRACT® DNA isolation kit provides a simple method to isolate DNA from plasma, serum, and body fluids.
- Rapid, provide results in less than 15 minutes
- Safe to use, contains only non-toxic reagents and no phenol chloroform
- Highly efficient, isolate DNA from plasma or serum containing as low as 0,1 ng DNA
: Supplied with digestion solution, digestion powder, DNA isolation buffer, DNA elution solution, spin column and collection tube.
Spécifications
Spécifications
- Cdt:1 KIT
- Désignation:EPIXTRACT® DNA isolation kit
- Cat. No.:ENZOENZ450180100
- Prep size:Mini
- Refine target molecule:Total DNA
- Target molecule:DNA
- Method / Format:Column (silica-based)
- Sample type:Biofluids (serum, plasma, saliva)
- Désignation:EPIXTRACT® DNA isolation kit



