Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

DNA isolation kit, EPIXTRACT®

EPIXTRACT® DNA isolation kit, 1 KIT
N° de catalogue ENZOENZ450180050
Fournisseur:   Enzo Life Sciences
DNA isolation kit, EPIXTRACT®
N° de catalogue ENZOENZ450180050
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
DNA isolation kit, EPIXTRACT®

EPIXTRACT® DNA isolation kit, 1 KIT

Numéro CAS:  
Prix
SFr. 301.00
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents

EPIXTRACT® DNA isolation kit provides a simple method to isolate DNA from plasma, serum, and body fluids.

  • Rapid, provide results in less than 15 minutes
  • Safe to use, contains only non-toxic reagents and no phenol chloroform
  • Highly efficient, isolate DNA from plasma or serum containing as low as 0,1 ng DNA

 : Supplied with digestion solution, digestion powder, DNA isolation buffer, DNA elution solution, spin column and collection tube.

Spécifications

Spécifications

  • Cdt:
    1 KIT
  • Désignation:
    EPIXTRACT® DNA isolation kit
  • Cat. No.:
    ENZOENZ450180050
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Biofluids (serum, plasma, saliva)
  • Désignation:
    EPIXTRACT® DNA isolation kit

Spécifications