Objectives for Rebel Microscopes
Achromat phase RNG-10/20 N.A. 0,40, 20×, 1,5 mm
Catalog # ECHLRMAG02033
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Specifications
- Grossissement:20×
- Désignation:Achromat phase RNG-10/20 N.A. 0,40
- Seq:15
- Distance de travail:1,5 mm