[EN]BOTTLE PP 125ML ROUND NM BATCH CONTR 1 * 50 Pce
Catalog # VWRL2153428S
Supplier: LAB3 OWN PRODUCTION
[EN]BOTTLE PP 125ML ROUND NM BATCH CONTR 1 * 50 Pce
Catalog # VWRL2153428S
Supplier: LAB3 OWN PRODUCTION
Frequently Bought Together
RAKO BEHÄLTER GRUNDFLÄCHE BLAU 600 X 400 X 425 MM 1 * 1 ST
Supplier: PLASTIC HAUS
VWR®, Capsules, évaporation, forme haute
Supplier: VWR Collection
Béchers, forme basse
Supplier: Bohlender
MultiRack racked tips, Axygen®
Supplier: Corning
Jerricans, narrow neck, HDPE, UN Approved
Supplier: LAMAPLAST
[EN]CLOSURE DIN60 BLUE WITH CONE 1 * 300 Pce
Supplier: MAUSER
Pissettes à col étroit, avec bouchon à vis
Supplier: Brand
Multiple distributors for bottles , BOLA
Supplier: Bohlender
Buchner Table-Top Funnels, 610 mm (24"), SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Corning® DeckWorks Reload Systems
Supplier: Corning
SERVI-MEDICAL Wastel 60 l blau ohne Aufkleber 060V4080BL 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Fer (II) sulfate heptahydraté ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals