ContactUsLinkComponent
RNAscope® HiPlex Probe Diluent - RNAscope® HiPlex Probe Diluent diluent is used to dilute 50x target probes to 1X before probe hybr idization step. It can also be used as a No Probe negative control. 1 * 1 ST
Référence ADVD324301
RNAscope® HiPlex Probe Diluent - RNAscope® HiPlex Probe Diluent diluent is used to dilute 50x target probes to 1X before probe hybr idization step. It can also be used as a No Probe negative control. 1 * 1 ST
Référence ADVD324301
Frequently Bought Together