Codesoft Lite Software
Codesoft lite subscription USB
Catalog # BRDY196184
Supplier: Brady
Jump to:
Product Details & Documents
Codesoft Lite label design software can be used for asset tracking, barcode labelling, and inventory and inspection labelling applications.
- Only available as a single user license
- Limited to 1 thermal printer
Languages availble include Arabic (Egypt), Czech, Danish, Dutch, English, Finnish, French, German, Hebrew, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Spanish, Swedish, and Turkish.
Compatible with BBP11, BBP12, BBP72, PR Plus, IP Series, PAM3000, PAM3600, THT 76mm Core, and TLS-PC LINK printers.
Specifications
- Désignation:Codesoft lite subscription USB
- Seq:02
- Cat. No.:BRDY196184