,,7 components: 50mg/l each of F- Cl- NO2- Br- NO3- PO43- SO42- in H2O ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months. (6 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
N° de catalogue CMPLRM044557L1
Fournisseur: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together