ContactUsLinkComponent
,,22 components: Al 500mg/l As 50mg/l Ba 50mg/l Bi 50mg/l Cd 50mg/l Co 50mg/l Cr 50mg/l Hg 5mg/l Mo 50mg/l Ni 50mg/l Pb 50mg/l Sb 50mg/l Se 50mg/l Sn 50mg/l Sr 50mg/l Tl 50mg/l V 50mg/l Te 50mg/l Cu 500mg/l Fe 50 1 * 100 mL
Catalog # CMPLRM018678L1
,,22 components: Al 500mg/l As 50mg/l Ba 50mg/l Bi 50mg/l Cd 50mg/l Co 50mg/l Cr 50mg/l Hg 5mg/l Mo 50mg/l Ni 50mg/l Pb 50mg/l Sb 50mg/l Se 50mg/l Sn 50mg/l Sr 50mg/l Tl 50mg/l V 50mg/l Te 50mg/l Cu 500mg/l Fe 50 1 * 100 mL
Catalog # CMPLRM018678L1
Frequently Bought Together