Self-laminating Polyester Labels for M710
Labels, rectangular, B-461, M7-115-461, Polyester, Permanent, acrylique, White/Transparent, M710, 25,40W×66,68H mm, 100/box
N° de catalogue BRDY311976
Fournisseur: Brady
Aller à
Product Details & Documents
Self-laminating polyester labels for M710 printer.
- Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
- Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
Spécifications
- Imprimeur:M710
- Type:B-461
- Couleur:White/Transparent
- Désignation:Labels, rectangular
- Dimensions:25,40W×66,68H mm
- Matière:Polyester
- Seq:07
- Modèle:M7-115-461
- Emballage:100/box
- Adhésif:Permanent, acrylique