High Sensitivity RNA ScreenTape Analysis
Reliable, reproducible, and objective measurements of RNA samples of limited abundance.
- Utilises RINe – a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
- Fast results are obtained in approximately 2 minutes per sample
- Minimal sample consumption is required as only 2 µl of sample is needed
- Detection of RNA sample with limited abundance down to 100 pg/µl is possible
- Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR workflows
- Ready-to-use consumables provide excellent reproducibility
- No cross contamination and sample carryover by using single lanes and individual loading tips
- Partially used ScreenTape devices can be utilised for completion at a later point in time
High Sensitivity RNA ScreenTape analysis provides efficient and reliable separation of total RNA samples of limited abundance down to 100 pg/µl. Compatible with eukaryotic and prokaryotic samples, the assay provides quality, quantity, and sizing information. Objective evaluation of RNA degradation is delivered with an RNA integrity number (RINe).
Results are obtained quickly, as fast as 2 minutes per sample. Accurate analysis is possible with only 2 µl of sample.
The assay is compatible with all Agilent TapeStation instruments.
Note: Some products within this assay contain SYBR™ Gold, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
: For research use only. Not for use in diagnostic procedures.
- Désignation:High sensitivity RNA ScreenTape sample buffer
- Seq:03
- Details:For the high sensitivity analysis of total RNA down to a sensitivity of 100 pg/µl, sample buffer only, to order with AGLS5067-5579 and AGLS5067-5581.