5 components: Ammonium (NH4+) 1000mg/l Potassium (K+) 1000mg/l Sodium (Na+) 1000mg/l Lithium (Li+) 200mg/l Ethanolamine (for Inorganics) (C2H7NO) 1000mg/l in Water CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after openin 1 * 500 mL
Référence CMPLRM092685L5
Fournisseur: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together