SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
NI 1MG/L IN HNO3 2% 1 * 100 mL
CMPLRM001477L1
:
CUSTOM MADE CHEMICALS LAB
NI 1MG/L IN HNO3 2% 1 * 100 mL
CMPLRM001477L1
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
KIT REACTIFS AMTAX SC 1-100 mg/l NH4-N 1 * 1 KIT
: Hach
Sodium hydroxyde 30% dans l'eau, grade Technique
: Thermo Fisher Scientific
[EN]1 COMPONENT OS 1000MG/L IN HCL 2% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
[EN]6 COMPONENTS IN HNO3 2% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
Rouge huile O 0,5% dans le propylène glycol, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY