Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

ethanedithioamide 98%

Bottle
Catalog # APOSOR24680-1G
Irritant
Supplier:   Apollo Scientific
ethanedithioamide 98%
Catalog # APOSOR24680-1G
Supplier:  Apollo Scientific
CAS Number:  79-40-3
ethanedithioamide 98%

Bottle

CAS Number:  79-40-3
Price
SFr. 31.20
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Ethanebis(thioamide), Rubeanic acid
  • CAS Number:79-40-3
  • Formula:C₂H₄N₂S₂
  • MDL Number:MFCD00004941
  • Molecular Weight:120,19655 g/mol
  • EINECS:201-203-9
Specifications

Specifications

  • Type de conditionnement:
    Bottle

Specifications