Specifications
See all product configurations- Catalog No:
- MFLX78006-46-CH
- MFLX78006-26-EU
- MFLX78006-48-CH
- MFLX78006-22
- MFLX78006-44-CH
- MFLX78006-42-CH
- MFLX78006-28-CH
- MFLX78006-26-CH
- MFLX78006-24-CH
- MFLX78006-22-CH
- MFLX78006-28
- MFLX78006-26
- MFLX78006-24-EU
- MFLX78006-24
- MFLX78006-22-EU
- MFLX78006-48-EU
- MFLX78006-48
- MFLX78006-46-EU
- MFLX78006-46
- MFLX78006-44-EU
- MFLX78006-44
- MFLX78006-42-EU
- MFLX78006-42
- MFLX78006-28-EU
- Modèle:
- 78006-46
- 78006-26
- 78006-48
- 78006-22
- 78006-44
- 78006-42
- 78006-28
- 78006-26
- 78006-24
- 78006-22
- 78006-28
- 78006-26
- 78006-24
- 78006-24
- 78006-22
- 78006-48
- 78006-48
- 78006-46
- 78006-46
- 78006-44
- 78006-44
- 78006-42
- 78006-42
- 78006-28
- Nombre de canaux:
- 16
- 16
- 24
- 8
- 12
- 8
- 24
- 16
- 12
- 8
- 24
- 16
- 12
- 12
- 8
- 24
- 24
- 16
- 16
- 12
- 12
- 8
- 8
- 24
- Nombre de rouleaux:
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- 8
- Type de commande:
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Min. flow rate per channel:
- 0,0004 ml /min
- 0,002 ml/min
- 0,0004 ml /min
- 0,002 ml/min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,002 ml/min
- Max. flow rate per channel:
- 11 ml/min
- 44 ml/min
- 11 ml/min
- 44 ml/min
- 11 ml/min
- 11 ml/min
- 44 ml/min
- 44 ml/min
- 44 ml/min
- 44 ml/min
- 44 ml/min
- 44 ml/min
- 44 ml/min
- 44 ml/min
- 44 ml/min
- 11 ml/min
- 11 ml/min
- 11 ml/min
- 11 ml/min
- 11 ml/min
- 11 ml/min
- 11 ml/min
- 11 ml/min
- 44 ml/min
- Débit min.:
- 0,0004 ml /min
- 0,002 ml/min
- 0,0004 ml /min
- 0,002 ml/min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,002 ml/min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,0004 ml /min
- 0,002 ml/min
- Drive IP rating:
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- IP 31
- Précision:
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- ±1,0%
- Anti-goutte:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Connection Cloud:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Digital dispensing:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Entraînement:
- IPC 11
- IPC 45
- IPC 11
- IPC 45
- IPC 11
- IPC 11
- IPC 45
- IPC 45
- IPC 45
- IPC 45
- IPC 45
- IPC 45
- IPC 45
- IPC 45
- IPC 45
- IPC 11
- IPC 11
- IPC 11
- IPC 11
- IPC 11
- IPC 11
- IPC 11
- IPC 11
- IPC 45
- Duty cycle:
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Min. RPM:
- 0,11 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,11 rpm (min⁻¹)
- 0,4 rpm (min⁻¹)
- Open-head sensor:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Remote I/O:
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- Reversible motor:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Series:
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Tubing sizes accepted:
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Two-stop 0,13 - 3,17 mm
- Washdown capable:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Pression max.:
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- 1 bar (14,5 psi)
- Fréquence:
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- Tension:
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- Garantie:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- Currently Viewing
Specifications
About this item
Obtenez un débit multicanal de haute précision, avec une interface intuitive à écran tactile.
- Replicate same flow for all channels; perfect for auto-analysis, reactor studies, cell line seeding, and research
- Planetary drive with 8 stainless steel rollers reduces pulsation, increases flow rate accuracy, and lengthens tubing life
- 12,7 cm (5") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
- Flexible operational control via screen or remote control (analog remote control, serial, or EtherNet/IP)
- Three-level user access – gives you the option of controlling who can operate the pump and at what level
- Pump lets you save and retrieve commonly used dispense programs for rapid set up
- Continuous run, volume dispense, and time dispense modes; batch control and interval/off time in volume and time modes
These Ismatec® IPC multichannel pumps feature planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The intuitive touchscreen display shows flow rate or RPM (min⁻¹), allows for easy calibration, features configurable volume or time dispense modes, and lets you save and recall up to 50 programs. Volumetric dispensing features a unique roller positioning system to ensure repeatability.
Mix and match tubing sizes and formulations across the channels to suit your application. Included Click’n’go™ cartridges feature automatic occlusion for simplified use, fewer tubing changes, and reproducible results. Optional pressure lever cartridges offer you the opportunity to manually adjust the back pressure experienced by the tubing / pump combination (order separately).
Pumps feature a variety of built in options for remote communications. EtherNet/IP control is available via RJ45. RS-232 serial control is available via a DB9 interface or USB port (USB use requires cable MFLX78006-90). A 25-pin connector allows analog input and output controls.
- Puissance:90 - 260 VAC; 50/60 Hz
- Précision:±1,0%
- Connection Cloud:Yes
- Type de commande:Digital
- Fréquence:50/60 Hz
- Pression max.:1 bar (14,5 psi)
- Débit min.:0,0004 ml /min
- Nombre de canaux:16
- Tension:90 - 260 VAC
- Désignation:Masterflex® Ismatec® IPC peristaltic pump, 0.1 - 11.25 rpm, 16-channel, 115/230 VAC
- Hauteur:200 mm
- Longueur:430 mm
- Seq:22
- Environmentally Preferable:
- Modèle:78006-46
- Duty cycle:Continuous
- Anti-goutte:Yes
- Digital dispensing:Yes
- Drive IP rating:IP 31
- Max. flow rate per channel:11 ml/min
- Min. flow rate per channel:0,0004 ml /min
- Min. RPM:0,11 rpm (min⁻¹)
- Open-head sensor:No
- Remote I/O:EtherNet/IP, Serial RS-232, MasterflexLive, 0 - 20 mA, 4 - 20 mA, 0 - 10 V, start/stop, direction, and prime via 25 pin contact closure
- Reversible motor:Yes
- Tubing sizes accepted:Two-stop 0,13 - 3,17 mm
- Washdown capable:No
- Entraînement:IPC 11
- Nombre de rouleaux:8
- Garantie:2 years
- Series:Ismatec
- Cdt:1
- Largeur:193 mm
- Electrical plug type:CH