Pipette tips
Standard, 0,1 - 10 µl, -, Transparent, Gilson® P2, P10, Bulk
N° de catalogue KART0095600
Fournisseur: KARTELL
Aller à
Product Details & Documents
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- Tips in racks come with snap-on lid
- Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
- Containers and tips are autoclavable to +121 °C for 20 minutes
Spécifications
- Emballage:Bulk
- Stérile:-
- Volume:0,1 - 10 µl
- Seq:01
- Style de pointe:Standard
- Couleur de pointe:Transparent
- Pour l'utilisation avec:Gilson® P2, P10