ContactUsLinkComponent
AXON MEDCHEM CLK8 A SPECIFIC CLOCK-BINDING SMALL MOLECULE, INHIBITS THE INTERACTION BETWEEN CLOCK AND BMAL1 AND INTERFERES WITH NUCL EAR TRANSLOCATION OF CLOCK BOTH IN VIVO AND IN VITRO. CLK8 CAN BE USED TO ENHANCE CIRCADIAN RHYTHM. 1 * 5 mg
Référence MSPP3224-5MG
AXON MEDCHEM CLK8 A SPECIFIC CLOCK-BINDING SMALL MOLECULE, INHIBITS THE INTERACTION BETWEEN CLOCK AND BMAL1 AND INTERFERES WITH NUCL EAR TRANSLOCATION OF CLOCK BOTH IN VIVO AND IN VITRO. CLK8 CAN BE USED TO ENHANCE CIRCADIAN RHYTHM. 1 * 5 mg
Référence MSPP3224-5MG
Frequently Bought Together