ContactUsLinkComponent
RNASCOPE LS 2.5 4-PLEX POSITIVE CONTROL PROBE HS-RNASCOPE LS 2.5 HUMAN POSITIVE CONTROL PROBE FOR RNASCOPE LS 2.5 MULTPLEX FLUORESCE NT ASSAY POLR21 ( C1 CHANNEL) AND PPIB C2 CHANNEL UBC C3 CHANNEL AND HPRT-1 1 * 1 ST
Référence ADVD321808
RNASCOPE LS 2.5 4-PLEX POSITIVE CONTROL PROBE HS-RNASCOPE LS 2.5 HUMAN POSITIVE CONTROL PROBE FOR RNASCOPE LS 2.5 MULTPLEX FLUORESCE NT ASSAY POLR21 ( C1 CHANNEL) AND PPIB C2 CHANNEL UBC C3 CHANNEL AND HPRT-1 1 * 1 ST
Référence ADVD321808
Frequently Bought Together