FITC-Labeled Human CD98, His Tag (CD8-HF248) is expressed from human 293 cells (HEK293). It contains AA Arg 206 - Ala 630 (Accession # P08195-1). 1 * 200 µG
FITC-Labeled Human CD98, His Tag (CD8-HF248) is expressed from human 293 cells (HEK293). It contains AA Arg 206 - Ala 630 (Accession # P08195-1). 1 * 200 µG