Masterflex® Fitting, PVDF, Tee, Hosebarb to Thread Adapter, 3/8" ID x 3/8" ID x 3/8"NPT(M); 10/Pk
Catalog # MFLX40615-74
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Taille de l'ardillon/Tube Ø int. (port 2):3/8"
- Type de connection:Thread to Barb
- Flow pattern:T
- Désignation:Fitting, PVDF, Tee, Hosebarb to Thread Adapter, 3/8" ID x 3/8" ID x 3/8"NPT(M); 10/Pk
- Environmentally Preferable:
- Orifice dimensions:0,25"
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Cdt:10
- Barb size/tubing int.Ø (port 3):3/8"
- Fitting material:PVDF
Frequently Bought Together