Micropump L22059 Gear Pump Service Kit for 07003-02; 1/Ea
N° de catalogue MFLX07003-12
Fournisseur: Avantor Fluid Handling
À propos de cet article
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
- Désignation:Gear Pump Service Kit, for 07003-02
- Environmentally Preferable:
- Garantie:1 year
- Cdt:1
Frequently Bought Together