ContactUsLinkComponent
Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 Fitting
Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 Fitting
Codice catalogo MFLX78167-56
Nome del fornitore:  Avantor Fluid Handling
1 year warranty
undefined
Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 Fitting
Codice catalogo MFLX78167-56
Nome del fornitore:  Avantor Fluid Handling
1 year warranty
Informazioni su questo articolo
Replacement parts keep your dosing pump running smoothly.
  • Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
  • Désignation:
    Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 Fitting
  • Environmentally Preferable:
  • Wetted materials:
    FEP
  • Garantie:
    1 year
  • Cdt:
    1
Frequently Bought Together