GF Signet 3-9900 Batch Module for Flow Transmitter 56560-12
Catalog # MFLX56560-13
Supplier: Avantor Fluid Handling
About this item
Optimize performance of your GF+ Signet Transmitter/Controller.
- Choose from a wide range of replacement parts and accessories
Use modules, sensors, and RC filters to expand the controller's capabilities. Adapt your controller to your existing system with a variety of mounting options.
- Type de commande:On/Off
- Désignation:Batch Module for Flow Transmitter 56560-12
- Environmentally Preferable:
- Garantie:See message
- Cdt:1
Frequently Bought Together