Masterflex® Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 3/16" ID x 1/4 NPT(M); 100/PK
Catalog # MFLX31702-07
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/16" ID x 1/4 NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Taille de l'ardillon/Tube Ø int. (port 2):3/16"
- Type de connection:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1/4"
- Désignation:Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 3/16" ID x 1/4 NPT(M); 100/PK
- Seq:59
- Environmentally Preferable:
- Modèle:31702-07
- No. of ports:2
- Sterilisation:Ethylene oxide (ETO) only
- Garantie:1 year
- Cdt:100
- Fitting material:Nylon
Frequently Bought Together