ContactUsLinkComponent
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.100" Diameter
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.100" Diameter
Catalog # MFLX17315-21
undefined
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.100" Diameter
Catalog # MFLX17315-21
About this item
Mix, react, and/or separate liquid-liquid mixtures in a single device.
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.
  • Ø:
    2,794 cm
  • Désignation:
    Centrifugal Multi-Purpose Separator/Extractor Weir; 1,100" Diameter
  • Environmentally Preferable:
  • Garantie:
    2 years
  • Cdt:
    1
Frequently Bought Together