GF Signet P526-2010 316 SS Socket-Weld Mini-Tap Fittings; 1" Pipe ID
N° de catalogue MFLX05618-63
Fournisseur: Avantor Fluid Handling
À propos de cet article
Installation fittings for your GF® Signet Paddle Wheel Flowmeters.
- 316 SS Socket-Weld Mini-Tap Fittings for Metalex™ sensors only
- Includes cap kit
Fittings are precision crafted to ensure proper sensor insertion depth and accurate flow measurement. Pipe fitting sizes range from 1/2" to 1" in diameter.
Required system components are the flow sensor, installation fitting, and monitor or controller, all sold separately.
- Désignation:316 SS Socket-Weld Mini-Tap Fittings; 1" Pipe ID
- Environmentally Preferable:
- Wetted materials:316 Stainless Steel
- Garantie:1 year part and 2 years labor
- Cdt:1
- Pipe size:1"
Frequently Bought Together