Masterflex® L/S® High-Performance Pump Head Mounting Hardware, Avantor®
L/S® High-Performance Pump Head Replacement Mounting Kit. 2 years warranty, L/S
Catalog # MFLX77252-01
Supplier: Avantor Fluid Handling
Masterflex® L/S® High-Performance Pump Head Mounting Hardware, Avantor®
Catalog # MFLX77252-01
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Secure your Masterflex® High-Performance pump head!
- Constructed of durable stainless steel
Keep your pump head operating at its best with these replacement parts.
Delivery information: Includes four 10-24 stainless steel mounting screws.
Specifications
- Désignation:L/S® High-Performance Pump Head Replacement Mounting Kit. 2 years warranty
- Seq:02
- Pump series:L/S